Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ PETG Sterile Diagnostic Vials

Catalog No. 01670077
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01-670-077 20 mL
01-670-075 5 mL
01-670-076 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-670-077 Supplier Qorpak™ Supplier No. 334341
Add to Cart
Add to Cart

Leak-proof HDPE closures ensure safe storage and transport of biological materials like antibodies, proteins, nucleic acids, enzymes, substrates, reagents and buffers.

  • Ideal for clinical diagnostic laboratories
  • Crafted with precision, these vials are USP Class VI compliant, ensuring non-cytotoxic and non-pyrogenic properties for abnormal toxicity
  • Specifically designed to meet the critical requirements of in-vitro fertilization media and all cell culture fluids
  • Suitable for air shipment with each manufacturing run tested to guarantee leak-proof performance
  • Manufactured in ISO-approved cleanroom facility using USP Class VI raw material
  • Provides excellent transparency similar to glass
  • Features HDPE cap closure with polyethylene cap liner for secure sealing
  • Gamma sterile SAL 10-6 product ensures maximum sterility
  • Tested for low leachables and extractables
  • Non-pyrogenic and non-cytotoxic for safe sample handling
  • Tested to ensure consistent leak-proof performance

Specifications

Closure Color White
Material (Liner) PE
Color Clear
Diameter (Metric) Outer 30 mm
Material PETG
Pre-slit No
Sterility Sterile
Disposable Single-use
Volume (Metric) 20 mL
Graduated No
Includes Closure
Neck Style Screw On
No. per Case 100
Base Style FLAT
Closure Material HDPE
Closure Type Screw Cap
Quantity 100
Closure Size 20-415
Marking Spot No
Fit for Purpose Sterile
Autoclavable Not Autoclavable
Certifications/Compliance USP, Class VI
Height (Metric) 65 mm
Preservative No
Product Type Vial
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.