Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Marketlab PETG Triple Glove Box Dispenser with Single Bag Holder

Catalog No. 22505175
Change view
Click to view available options
No. of Compartments:
4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Compartments
22-505-175 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-505-175 Supplier Marketlab Supplier No. PS0040111
Add to Cart
Add to Cart

Designed to organize gloves and bag rolls for convenient access in healthcare and clinical environments.

  • Holds three boxes of gloves and one roll of individual-sized biohazard, linen, or garbage bags
  • Two-way keyholes for wall mounting
  • Clear PETG plastic
  • Dispenses boxes no larger than 5.25 in.W x 10 in.H x 3.75 in.D and rolls no larger than 10 in.W x 3.4 in.
  • Clear material allows full view of box details
  • Mountable either vertically or horizontally to fit space constraints
  • Suggested cleaning method is to use a soft cloth, warm water, and mild soap

Specifications

Product Type Glove Dispenser
Color Clear
Depth (English) 3.95 in.
Width (English) 10.1 in.
Height (English) 20.4 in.
Holds Gloves
Material PETG
Mounting Keyhole Slots
No. of Compartments 4
Product Style Side Loading
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.