Learn More
Alkali Scientific Petri Dish 100X15mm, Sterile, Standard, Stackable, Semi Slippable, 20 per Inner Bag, 500/cs
SDP
Catalog No. 502336052
Encompass_Preferred
These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange and comparents for bacterial culture use with different media under the same conditions. 500/cs
Catalog No. 50-233-6052 Supplier Alkali Scientific Supplier No. PD3571
These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange and comparents for bacterial culture use with different media under the same conditions. 500/cs