Learn More
Medchemexpress LLC Petrelintide acetate | 99.8% | 4191.69 (free base) | 5 MG
SDP
Catalog No. 5000425654
Encompass_Preferred
Petrelintide acetate is a dual amylin and calcitonin receptor agonist (DACRA). It is a white to off-white solid with a molecular weight of 4191.69 (free base) and a chemical formula of C185H305N49O61.xC2H4O2. Primarily used for laboratory research and substance manufacturing, it is not classified as a hazardous substance.
  • Dual amylin and calcitonin receptor agonist activity
  • Selectively reduces high-fat diet intake in DIO rats
  • Promotes fat mass loss while preserving lean mass
  • Useful in diabetes research
  • Exhibits agonistic activity against hAMY3R and hCTR in HEK-293 cells
Catalog No. 50-004-25654 Supplier Medchemexpress LLC Supplier No. HYP10745A5MG
Petrelintide acetate is a dual amylin and calcitonin receptor agonist (DACRA). It is a white to off-white solid with a molecular weight of 4191.69 (free base) and a chemical formula of C185H305N49O61.xC2H4O2. Primarily used for laboratory research and substance manufacturing, it is not classified as a hazardous substance.
  • Dual amylin and calcitonin receptor agonist activity
  • Selectively reduces high-fat diet intake in DIO rats
  • Promotes fat mass loss while preserving lean mass
  • Useful in diabetes research
  • Exhibits agonistic activity against hAMY3R and hCTR in HEK-293 cells