Learn More
Electron Microscopy Sciences Petrographic Thin Section 25 Slide Storage Box, Black
SDP
Catalog No. 501931403
Encompass_Preferred
Designed for safely storing petrographic slides long term and also for transport. Made from durable ABS plastic with a hinged lid, secure latch, and foam base.
Color: Black
Catalog No. 50-193-1403 Supplier Electron Microscopy Sciences Supplier No. 7153405
Designed for safely storing petrographic slides long term and also for transport. Made from durable ABS plastic with a hinged lid, secure latch, and foam base.
Color: Black