Learn More
Chemglass Life Sciences Chemglass Petri Dish, Non-Treated, Sterile, 100 x 20 mm (Nest)
SDP
Catalog No. 502733503
Encompass_Preferred
Chemglass sterile, non-treated Petri dish manufactured from high-clarity polystyrene for general microbiology and culture applications. Dimensions are 100 x 20 mm; dish is non-pyrogenic and sterilized by electron beam.
  • High clarity polystyrene.
  • Dimensions: 100 x 20 mm.
  • Non-treated surface.
  • Sterilized by electron beam (E-Beam); SAL = 10^-6.
  • Non-pyrogenic.
  • PA/PE packaging.
  • Vent for gas exchange.
  • Numeric mark on bottom for identification.
  • Stackable for storage and handling.
Catalog No. 50-273-3503 Supplier Chemglass Life Sciences Supplier No. CGN1802101
Chemglass sterile, non-treated Petri dish manufactured from high-clarity polystyrene for general microbiology and culture applications. Dimensions are 100 x 20 mm; dish is non-pyrogenic and sterilized by electron beam.
  • High clarity polystyrene.
  • Dimensions: 100 x 20 mm.
  • Non-treated surface.
  • Sterilized by electron beam (E-Beam); SAL = 10^-6.
  • Non-pyrogenic.
  • PA/PE packaging.
  • Vent for gas exchange.
  • Numeric mark on bottom for identification.
  • Stackable for storage and handling.