Learn More
Andwin Scientific PETRI DISH 100X15MM CS500
SDP
Catalog No. NC0476581
Encompass_Preferred
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a stackable design that is useful for automation. These dishes are measured to ha
Catalog No. NC0476581 Supplier Andwin Scientific Supplier No. 25384302QT
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a stackable design that is useful for automation. These dishes are measured to ha