Learn More
Mascon Medical Mascon Medical™ Petri Dish, 100*20mm, Sterile 10EA/BAG, 50 Bags/ Case
SDP
Catalog No. NC2458345
Encompass_Preferred
The petri dish series products provided by Mascon Medical™ are made of high-quality medical-grade polystyrene with full specifications and high transparency. They are widely used in a variety of bacterial culture environment test lamp experiments.Features:•Meets ISO13485 quality management system standard•Stackable and easy to store•Nonpyrogenic•No endotoxin•RNase/DNase FreeSpecifications:•Volume: 100*20mm •Qty. / Case: 500•Material: Polystyrene•Sterile: Yes•Size: Φ98*17.4mm•Temperature Range: -40℃~100℃•Packaging Format: 10 EA/Bag, 50 Bags/Case
Catalog No. NC2458345 Supplier Mascon Medical Supplier No. FSPETRIDISH
The petri dish series products provided by Mascon Medical™ are made of high-quality medical-grade polystyrene with full specifications and high transparency. They are widely used in a variety of bacterial culture environment test lamp experiments.Features:•Meets ISO13485 quality management system standard•Stackable and easy to store•Nonpyrogenic•No endotoxin•RNase/DNase FreeSpecifications:•Volume: 100*20mm •Qty. / Case: 500•Material: Polystyrene•Sterile: Yes•Size: Φ98*17.4mm•Temperature Range: -40℃~100℃•Packaging Format: 10 EA/Bag, 50 Bags/Case