Learn More
Chemglass Life Sciences Petri Dish, 150 x 15mm, Stackable, Sterile, 10/Pack, 100/Case
SDP
Catalog No. 501897753
Encompass_Preferred
These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange.High clarity polystyreneFlat, transparent surfaceStackable for easy storage and handlingNon-treatedSterilized by E-BeamNon-pyrogenic
Catalog No. 50-189-7753 Supplier Chemglass Life Sciences Supplier No. CGN1802145
These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange.High clarity polystyreneFlat, transparent surfaceStackable for easy storage and handlingNon-treatedSterilized by E-BeamNon-pyrogenic