Learn More
Chemglass Life Sciences Chemglass Petri Dish, Non-Treated, Sterile, Polystyrene, 150 x 25 mm, Nest
SDP
Catalog No. 502733502
Encompass_Preferred
Chemglass non-treated sterile petri dish for routine microbiology and plating applications. Manufactured from high-clarity polystyrene and packaged in nests for easy handling and stacking.
  • Material: Polystyrene.
  • Dimensions: 150 x 25 mm.
  • Surface treatment: Non-treated.
  • Sterility: Sterile; E-beam sterilized.
  • DNase/RNase-free and non-pyrogenic.
  • Packaging: Nest; stackable with bottom ridge for handling.
  • Features: High clarity, flat transparent surface; three vents for gas exchange.
Catalog No. 50-273-3502 Supplier Chemglass Life Sciences Supplier No. CGN1802151
Chemglass non-treated sterile petri dish for routine microbiology and plating applications. Manufactured from high-clarity polystyrene and packaged in nests for easy handling and stacking.
  • Material: Polystyrene.
  • Dimensions: 150 x 25 mm.
  • Surface treatment: Non-treated.
  • Sterility: Sterile; E-beam sterilized.
  • DNase/RNase-free and non-pyrogenic.
  • Packaging: Nest; stackable with bottom ridge for handling.
  • Features: High clarity, flat transparent surface; three vents for gas exchange.