Learn More
Andwin Scientific PETRI DISH 150X15MM CS100
SDP
Catalog No. NC0476610
Encompass_Preferred
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a stackable design that is useful for automation. These dishes are measured to ha
Catalog No. NC0476610 Supplier Andwin Scientific Supplier No. 25384326
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a stackable design that is useful for automation. These dishes are measured to ha