Learn More
TWD Scientific LLC Petri Dish, Twdbiox, 150X15Mm, Sterile, Polystyrene, Triple-Vented Lid, 200/Case.
SDP
Catalog No. 502285036 Shop All TWD Scientific LLC Products
Encompass_Preferred
150X15Mm Clear Polystyrene Sterile Petri Dish With Triple-Vented Lid. Petri Dishes Feature Stackable Design. 10/Pack, 20 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5036 Supplier TWD Scientific LLC Supplier No. BX661515
150X15Mm Clear Polystyrene Sterile Petri Dish With Triple-Vented Lid. Petri Dishes Feature Stackable Design. 10/Pack, 20 Packs/Case, Price/Case