Learn More
Chemglass Life Sciences Petri Dish, 2 Compartments, Non-Treated Sterile Polystyrene
SDP
Catalog No. 502733287
Encompass_Preferred
This sterile, non-treated Petri dish is molded from high-clarity polystyrene and features two compartments for simultaneous culture of different media. It is sterilized by e-beam, DNase/RNase-free, and non-pyrogenic; stackable for easy storage and handling.
  • Made from high-clarity polystyrene.
  • Non-treated surface for general microbiology use.
  • Sterilized by e-beam and DNase/RNase-free.
  • Two compartments allow separate cultures under the same conditions.
  • Three vents for gas exchange.
  • Stackable design for compact storage and easy handling.
  • Sold 20 per sleeve; larger case quantities available.
Catalog No. 50-273-3287 Supplier Chemglass Life Sciences Supplier No. CLS180402
This sterile, non-treated Petri dish is molded from high-clarity polystyrene and features two compartments for simultaneous culture of different media. It is sterilized by e-beam, DNase/RNase-free, and non-pyrogenic; stackable for easy storage and handling.
  • Made from high-clarity polystyrene.
  • Non-treated surface for general microbiology use.
  • Sterilized by e-beam and DNase/RNase-free.
  • Two compartments allow separate cultures under the same conditions.
  • Three vents for gas exchange.
  • Stackable design for compact storage and easy handling.
  • Sold 20 per sleeve; larger case quantities available.