Learn More
Chemglass Life Sciences Chemglass Petri Dish, 3 compartments, non-treated, sterile, 100 mm, polystyrene (pack of 20)
SDP
Catalog No. 502733286
Encompass_Preferred
Three-compartment, 100 mm sterile Petri dishes made from high-clarity polystyrene for plating multiple media under identical incubation conditions. The non-treated surface is not intended for tissue culture; dishes are supplied sterile and are DNase/RNase-free.
  • Diameter: 100 mm.
  • Compartments: 3.
  • Material: High-clarity polystyrene.
  • Sterilization: E-beam sterilized.
  • Packaging: 20 per sleeve; larger case quantities available.
  • Feature: Flat, transparent surface.
  • Feature: Stackable for easy storage and handling.
  • Feature: Non-pyrogenic and DNase/RNase-free.
  • Feature: Three vents for gas exchange.
Catalog No. 50-273-3286 Supplier Chemglass Life Sciences Supplier No. CLS180403
Three-compartment, 100 mm sterile Petri dishes made from high-clarity polystyrene for plating multiple media under identical incubation conditions. The non-treated surface is not intended for tissue culture; dishes are supplied sterile and are DNase/RNase-free.
  • Diameter: 100 mm.
  • Compartments: 3.
  • Material: High-clarity polystyrene.
  • Sterilization: E-beam sterilized.
  • Packaging: 20 per sleeve; larger case quantities available.
  • Feature: Flat, transparent surface.
  • Feature: Stackable for easy storage and handling.
  • Feature: Non-pyrogenic and DNase/RNase-free.
  • Feature: Three vents for gas exchange.