Learn More
Chemglass Life Sciences Petri Dish, 35 x 12mm, Sterile, 20/Bag, 500/Case
SDP
Catalog No. 501897751
Encompass_Preferred
These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange.High clarity polystyreneFlat, transparent surfaceStackable for easy storage and handlingNon-treatedSterilized by E-BeamNon-pyrogenic
Catalog No. 50-189-7751 Supplier Chemglass Life Sciences Supplier No. CGN1802035
These non-treated polystyrene plates are transparent and DNase/RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange.High clarity polystyreneFlat, transparent surfaceStackable for easy storage and handlingNon-treatedSterilized by E-BeamNon-pyrogenic