Learn More
Chemglass Life Sciences Petri Dish, 4 Compartments, 100 mm, Non-Treated, Sterile Polystyrene
SDP
Catalog No. 502733285
Encompass_Preferred
Clear 100 mm polystyrene petri dish with four isolated compartments for parallel culture experiments. Sterile and non-treated, the plate features vents for gas exchange and a stackable design for easy storage.
  • 100 mm diameter.
  • High clarity polystyrene.
  • Sterile and non-treated surface.
  • Sterilized by E-beam.
  • DNase/RNase-free and non-pyrogenic.
  • Molded compartments allow separate cultures under the same conditions.
  • Vents for gas exchange.
  • Packaged 20 per sleeve; larger case quantities available.
Catalog No. 50-273-3285 Supplier Chemglass Life Sciences Supplier No. CLS180404
Clear 100 mm polystyrene petri dish with four isolated compartments for parallel culture experiments. Sterile and non-treated, the plate features vents for gas exchange and a stackable design for easy storage.
  • 100 mm diameter.
  • High clarity polystyrene.
  • Sterile and non-treated surface.
  • Sterilized by E-beam.
  • DNase/RNase-free and non-pyrogenic.
  • Molded compartments allow separate cultures under the same conditions.
  • Vents for gas exchange.
  • Packaged 20 per sleeve; larger case quantities available.