Learn More
Alkali Scientific Petri Dish, 60 x 12 mm, Non Vented for Anaerobic Applications, Sterile 20/Pk, 500/case
SDP
Catalog No. 5000662263
Encompass_Preferred
Sterile non vented Petri dishes measuring 60 × 12 mm are designed for anaerobic and microaerophilic culturing applications where maintaining a closed incubation environment is essential. The tight fitting lid minimizes gas exchange, helping preserve anaerobic conditions and reducing exposure to oxygen during microbial incubation. Manufactured from optically clear virgin polystyrene, these dishes provide excellent transparency for colony observation and microscopic evaluation. The consistently flat bottom surface supports uniform media distribution and reliable microbial growth patterns. Each lot is sterilized using E Beam technology and certified non pyrogenic to ensure contamination free laboratory performance. The dishes are stackable for convenient storage and automated handling, making them suitable for microbiology laboratories conducting anaerobic culture studies, research experiments, and routine microbial analysis.
Catalog No. 50-006-62263 Supplier Alkali Scientific Supplier No. PD3562NV
Sterile non vented Petri dishes measuring 60 × 12 mm are designed for anaerobic and microaerophilic culturing applications where maintaining a closed incubation environment is essential. The tight fitting lid minimizes gas exchange, helping preserve anaerobic conditions and reducing exposure to oxygen during microbial incubation. Manufactured from optically clear virgin polystyrene, these dishes provide excellent transparency for colony observation and microscopic evaluation. The consistently flat bottom surface supports uniform media distribution and reliable microbial growth patterns. Each lot is sterilized using E Beam technology and certified non pyrogenic to ensure contamination free laboratory performance. The dishes are stackable for convenient storage and automated handling, making them suitable for microbiology laboratories conducting anaerobic culture studies, research experiments, and routine microbial analysis.