Learn More
Research Products International Corp Disposable Petri Dishes, Sterile, Stackable, 60 x 15 mm, 20 per Sleeve, 500 per Case
SDP
Catalog No. 501367529
Encompass
Convenient 60 mm diameter size utilizes less media per dish and exceptionally flat bottom ensures even distribution. Dish is manufactured from medical grade polystyrene with optically clear top and bottom viewing surfaces. Molded in ventilation ribs promote air circulation and reduce condensation during incubation procedures. Lid is designed for easy removal using one gloved hand.
Dishes are supplied sterile and packaged in sealed sleeves containing 20 dishes per sleeve and 500 dishes per case.
Optically clear polystyrene
Sterile
Disposable
Stackable
Catalog No. 50-136-7529 Supplier Research Products International Corp Supplier No. 162901
Convenient 60 mm diameter size utilizes less media per dish and exceptionally flat bottom ensures even distribution. Dish is manufactured from medical grade polystyrene with optically clear top and bottom viewing surfaces. Molded in ventilation ribs promote air circulation and reduce condensation during incubation procedures. Lid is designed for easy removal using one gloved hand.
Dishes are supplied sterile and packaged in sealed sleeves containing 20 dishes per sleeve and 500 dishes per case.
Optically clear polystyrene
Sterile
Disposable
Stackable