Learn More
Ted Pella Inc Polystyrene Petri Dishes, 60mm OD x 13.7mm, 10/pk
SDP
Catalog No. NC1667321
Encompass
Specialty size for use with McIlwain Tissue Chopper, but excellent for general laboratory use. Bottom outside dia. 60mm x 13.7mm height; Top outside dia. 63mm x 8.5mm height.
Catalog No. NC1667321 Supplier Ted Pella Inc Supplier No. 1018404
Specialty size for use with McIlwain Tissue Chopper, but excellent for general laboratory use. Bottom outside dia. 60mm x 13.7mm height; Top outside dia. 63mm x 8.5mm height.