Learn More
Alkali Scientific Petri Dish, 75 x 50mm, Rectangular Plate, Sterile, Anaerobic, with Pill Pocket for CO2 Tablet, 500/CS
SDP
Catalog No. 5000662264
Encompass_Preferred
Petri Dish, 75 x 50mm, Rectangular Plate, Anaerobic, with Pill Pocket for CO₂ Tablet, is designed for anaerobic culture applications, this 75 x 50mm rectangular Petri dish features a convenient integrated pill pocket to securely hold a CO₂ generating tablet. The innovative design ensures optimal gas diffusion for creating and maintaining an anaerobic environment, supporting the growth of oxygen sensitive microorganisms. Made from high quality, optically clear polystyrene, these dishes allow for easy sample observation and consistent results. Each dish is precision molded for uniform thickness and reliable stacking. Supplied 500 per case for efficient laboratory use. Key Features Anaerobic Design: Built in pill pocket accommodates a CO₂ generating tablet to support anaerobic culture conditions. Rectangular Format: 75 x 50mm plate offers an efficient layout for multiple samples or comparative studies. Optically Clear Polystyrene: Ensures excellent visibility for colony observation.
Catalog No. 50-006-62264 Supplier Alkali Scientific Supplier No. PD7550
Petri Dish, 75 x 50mm, Rectangular Plate, Anaerobic, with Pill Pocket for CO₂ Tablet, is designed for anaerobic culture applications, this 75 x 50mm rectangular Petri dish features a convenient integrated pill pocket to securely hold a CO₂ generating tablet. The innovative design ensures optimal gas diffusion for creating and maintaining an anaerobic environment, supporting the growth of oxygen sensitive microorganisms. Made from high quality, optically clear polystyrene, these dishes allow for easy sample observation and consistent results. Each dish is precision molded for uniform thickness and reliable stacking. Supplied 500 per case for efficient laboratory use. Key Features Anaerobic Design: Built in pill pocket accommodates a CO₂ generating tablet to support anaerobic culture conditions. Rectangular Format: 75 x 50mm plate offers an efficient layout for multiple samples or comparative studies. Optically Clear Polystyrene: Ensures excellent visibility for colony observation.