Learn More
Chemglass Life Sciences Chemglass Petri Dish, Non-Treated, Sterile, 90 x 15 mm, Safety Lock (Nest)
SDP
Catalog No. 502733504
Encompass_Preferred
The Chemglass 90 x 15 mm non-treated sterile Petri dish features a safety-lock lid and high-clarity polystyrene construction, designed for general microbiology and culture applications.
  • 90 x 15 mm dimensions
  • Virgin polystyrene, high clarity
  • Non-treated, hydrophobic surface that discourages cell attachment
  • Sterile; sterilized by e-beam or gamma irradiation (SAL 10^-6)
  • Safety-lock twist closure for secure seating
  • Molded lid spacers for gas exchange
  • Nest design; stackable for compact storage and handling
  • Numeric mark on bottom for easy identification
Catalog No. 50-273-3504 Supplier Chemglass Life Sciences Supplier No. CGN1802091
The Chemglass 90 x 15 mm non-treated sterile Petri dish features a safety-lock lid and high-clarity polystyrene construction, designed for general microbiology and culture applications.
  • 90 x 15 mm dimensions
  • Virgin polystyrene, high clarity
  • Non-treated, hydrophobic surface that discourages cell attachment
  • Sterile; sterilized by e-beam or gamma irradiation (SAL 10^-6)
  • Safety-lock twist closure for secure seating
  • Molded lid spacers for gas exchange
  • Nest design; stackable for compact storage and handling
  • Numeric mark on bottom for easy identification