Learn More
Tisch Scientific Petri Dishes with aluminum lids. Set of 12
SDP
Catalog No. NC1811935
Encompass
Petri Dishes with aluminum lids. Set of 12. Used with Andersen style viable impactors.
Catalog No. NC1811935 Supplier Tisch Scientific Supplier No. TE10704
Petri Dishes with aluminum lids. Set of 12. Used with Andersen style viable impactors.