Learn More
Electron Microscopy Sciences Petri Dish; Analyslide, 100 per pack
SDP
Catalog No. 50930428
Encompass_Preferred
The analyslide is like a petri dish on a slide. Use it as an examination and storage container for 47 mm filters. Its rectangular base fits any microscope stage; an inner cover ring offers airtight security for the samples and it does not interfere with viewing. It is also ideal for drying coated grids and absorbing water from grids that are supporting sections.
Catalog No. 50-930-428 Supplier Electron Microscopy Sciences Supplier No. 70650
The analyslide is like a petri dish on a slide. Use it as an examination and storage container for 47 mm filters. Its rectangular base fits any microscope stage; an inner cover ring offers airtight security for the samples and it does not interfere with viewing. It is also ideal for drying coated grids and absorbing water from grids that are supporting sections.