Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Petri Dish Caddy - Six Pocket
SDP

Catalog No. 22022541
Change view
Click to view available options
Positions:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
22-022-541 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-022-541 Supplier Poltex Supplier No. PTRCAD6
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organize your petri dishes in this benchtop organizer and caddy. Handles for carrying. Stack your items vertically to free up valuable space.

  • Non-slip rubber bumpons on base
  • Handles for carrying
  • Stack your items vertically to free up valuable space
  • Constructed from super durable, cleanable PETG

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Petri Dishes
Material PETG
Positions 1
Shape Rectangular
Stackable No
Sterility Non-sterile
Length (Metric) 215.9 mm
Includes Caddy only
Product Type Petri Dish Holder
Height (Metric) 233.68 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.