Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Petri Dish Dispenser Acrylic

Catalog No. 14127660 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-127-660 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-127-660
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acrylic petri dish dispenser to keep dishes organized and within reach.

  • Holds up to 30 x 100 mm Petri dishes
  • Two partitions for easy dish access and organization
  • Durable, clear acrylic for visibility and strength
  • Suitable for benchtop use or wall mounting
  • Mounting screws included for secure installation

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Petri Dishes (100 mm)
Material Acrylic
Positions 30
Shape Rectangular
Stackable No
Sterility Non-sterile
Mounting Counter or wall mount with included hardware
Dimensions (L x W x H) 8.7 x 4.2 x 9.3 in.
Product Type Petri Dish Holder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.