Learn More
Research Products International Corp Petri Dish Incubator Trays, Holds 5 Dishes, 9 7/8 x 9 5/16 x 3/8 Inch Tray, 3 Trays per Package
SDP
Catalog No. 50212617
Encompass
Use our stackable polypropylene trays to increase capacity of an incubator or for extra benchtop workspace. Perforated trays allow uniform circulation of CO2 inside the incubator. Designed to hold 5 x 100 mm or 15 x 60 mm diameter petri dishes. Trays have two large surface areas for marking. Polypropylene plastic; can be disinfected with 70% ethanol or common laboratory disinfectants.
Catalog No. 50-212-617 Supplier Research Products International Corp Supplier No. 248846
Use our stackable polypropylene trays to increase capacity of an incubator or for extra benchtop workspace. Perforated trays allow uniform circulation of CO2 inside the incubator. Designed to hold 5 x 100 mm or 15 x 60 mm diameter petri dishes. Trays have two large surface areas for marking. Polypropylene plastic; can be disinfected with 70% ethanol or common laboratory disinfectants.