Learn More
Cole-Parmer Chemware D1069545 PFA Petri Dish, 100 x 15 mm, 100 mL, 1/Pk
Catalog No. NC0708878 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for contamination-free biological applications. Features an inert, nonstick, noncontaminating surface. Lids not included. Autoclavable.
Catalog No. NC0708878 Supplier Cole-Parmer Supplier No. 0614802
Ideal for contamination-free biological applications. Features an inert, nonstick, noncontaminating surface. Lids not included. Autoclavable.