Learn More
Research Products International Corp Petri Dish Shrink Seals, 92 mm x 25 mm, Clear Color, 400 per Package
SDP
Catalog No. 50998464
Encompass
Shrink Seals are colorless transparent cellulose bands for sealing Petri dishes and test tubes. This product is fantastic when preparing in-house plates that are not used quickly. Ideal when dealing with sensitivity plates for TB and fungus cultures that require prolonged incubation. Easy to apply. The size of the standard band is 92mm x 25mm
Shrink seal is both bactericidal and fungicidal.
Catalog No. 50-998-464 Supplier Research Products International Corp Supplier No. 160262
Shrink Seals are colorless transparent cellulose bands for sealing Petri dishes and test tubes. This product is fantastic when preparing in-house plates that are not used quickly. Ideal when dealing with sensitivity plates for TB and fungus cultures that require prolonged incubation. Easy to apply. The size of the standard band is 92mm x 25mm
Shrink seal is both bactericidal and fungicidal.