Learn More
Chemglass Life Sciences Petri Dish, 100 x 15mm, Non Treated, Sterile
SDP
Catalog No. 501897569
Encompass
These non-treated polystyrene plates are transparent and DNase / RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange.
Catalog No. 50-189-7569 Supplier Chemglass Life Sciences Supplier No. CGE1802102
These non-treated polystyrene plates are transparent and DNase / RNase-Free. The bottom has a ridge allowing for easy handling and stacking. The plate has three vents for gas exchange.