Learn More
Electron Microscopy Sciences Petri Dish With Absorbent Pads
SDP
Catalog No. 501899356
Encompass_Preferred
A 50x9 mm petri dish design that is easily opened with one hand, yet closes to a tight seal by its unique double rim. Specimen samples are preserved until you are ready to analyze them. Humidity remains locked in. When loaded with a 47 mm absorbent pad, it is ideal for drying coated grids, removing water from grids that are supporting sections. Dishes sterilized by gamma radiation.
Catalog No. 50-189-9356 Supplier Electron Microscopy Sciences Supplier No. 70662
A 50x9 mm petri dish design that is easily opened with one hand, yet closes to a tight seal by its unique double rim. Specimen samples are preserved until you are ready to analyze them. Humidity remains locked in. When loaded with a 47 mm absorbent pad, it is ideal for drying coated grids, removing water from grids that are supporting sections. Dishes sterilized by gamma radiation.