Learn More
Electron Microscopy Sciences Petri Dishes; Open with One Hand Without Absorbent pads
SDP
Catalog No. 50930429
Encompass_Preferred
A 50x9 mm petri dish design that is easily opened with one hand, yet closes to a tight seal by its unique double rim. Specimen samples are preserved until you are ready to analyze them. Humidity remains locked in. When loaded with a 47 mm absorbent pad, it is ideal for drying coated grids, removing water from grids that are supporting sections. Dishes sterilized by gamma radiation.
Catalog No. 50-930-429 Supplier Electron Microscopy Sciences Supplier No. 70660
A 50x9 mm petri dish design that is easily opened with one hand, yet closes to a tight seal by its unique double rim. Specimen samples are preserved until you are ready to analyze them. Humidity remains locked in. When loaded with a 47 mm absorbent pad, it is ideal for drying coated grids, removing water from grids that are supporting sections. Dishes sterilized by gamma radiation.