Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Petri dish w/pad 47 mm 150/pk

Catalog No. 5000207741 Shop All Hach Company Products
Encompass

Petri dish with pad (Millipore)-47 mm-150/pk

Catalog No. 50-002-07741 Supplier Hach Company Supplier No. 2936300
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Petri dish with pad (Millipore)-47 mm-150/pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.