Learn More
Advantec MFS, Inc PETRI DISH W/PAD STERILE 100PK
Catalog No. NC0356223
Encompass_Preferred
Polystyrene Dishes with Absorbant Pad for culturing microorganisms on 47mm membrane filter. Standard size of 50 x 11 mm fit commercial incubators. Raised ridge for handling/secure stacking. Gama arrayed. 5 sleeves of 20. (500pk avail.)
Catalog No. NC0356223 Supplier Advantec MFS, Inc Supplier No. 42004A01
Polystyrene Dishes with Absorbant Pad for culturing microorganisms on 47mm membrane filter. Standard size of 50 x 11 mm fit commercial incubators. Raised ridge for handling/secure stacking. Gama arrayed. 5 sleeves of 20. (500pk avail.)