Learn More
Alkali Scientific ASI™ Petri Dish, Y-Plate (3-Section), 100 x 15mm, Stackable, Sterile, 20/pk, 500/cs
SDP
Catalog No. 503029264
Encompass_Preferred
Disposable Petri Dishes are easy to use and is efficient for microbial culture. High clarity polystyrene Petri dishes available in traditional 1 compartment styles as well as 2 compartment, 3 compartment, and 4 compartment options. Alkali Scientic Petri Dishes are designed to be used for bacterial cell culture applications in all types of labs. High clarity polystyrene Flat transparent surface Numeric mark on the bottom for easy identification PA/PE packaging Vent for gas exchange Stackable for easy storage and handling Non-pyrogenic Sterilized by E-Beam, SAL=10-6 Packing: 20/pk, 500/cs
Catalog No. 50-302-9264 Supplier Alkali Scientific Supplier No. PD3571Y
Disposable Petri Dishes are easy to use and is efficient for microbial culture. High clarity polystyrene Petri dishes available in traditional 1 compartment styles as well as 2 compartment, 3 compartment, and 4 compartment options. Alkali Scientic Petri Dishes are designed to be used for bacterial cell culture applications in all types of labs. High clarity polystyrene Flat transparent surface Numeric mark on the bottom for easy identification PA/PE packaging Vent for gas exchange Stackable for easy storage and handling Non-pyrogenic Sterilized by E-Beam, SAL=10-6 Packing: 20/pk, 500/cs