Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Petri Dish, 100 x 15mm, Non Treated, Sterile
SDP

Catalog No. NC1012109
Encompass_Preferred

Petri Dish, 100mm, Non Treated, Sterile, Case of 500

Catalog No. NC1012109 Supplier Chemglass Life Sciences Supplier No. CLS1802100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Petri Dish, 100mm, Non Treated, Sterile, Case of 500

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.