Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/cs

Catalog No. NC0731514 Shop All Cole-Parmer Products
Encompass_Preferred

Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/cs.

Catalog No. NC0731514 Supplier Cole-Parmer Supplier No. 0613900
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile Petri Dishes; 100 mm dia x 15 mm H; 500/cs.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.