Learn More
Research Products International Corp Disposable Petri Dishes, Sterile, 150 x 15 mm, 10 per Sleeve, 100 per Case
SDP
Catalog No. 501367653
Encompass
Ideal optically clear petri dishes for automation and general microbiological use. Dishes are molded from virgin polystyrene and are radiation sterilized. Feature ventilation ribs for free air circulation and reduce condensation during incubation. Flared lid skirt and squared corners allow easy one hand operation.
Catalog No. 50-136-7653 Supplier Research Products International Corp Supplier No. 160215
Ideal optically clear petri dishes for automation and general microbiological use. Dishes are molded from virgin polystyrene and are radiation sterilized. Feature ventilation ribs for free air circulation and reduce condensation during incubation. Flared lid skirt and squared corners allow easy one hand operation.