Learn More
Andwin Scientific PETRI DISHES 100X15MM 600/CS
SDP
Catalog No. NC0788436
Encompass_Preferred
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a stackable design that is useful for automation. These dishes are measured to ha
Catalog No. NC0788436 Supplier Andwin Scientific Supplier No. 25384094
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a stackable design that is useful for automation. These dishes are measured to ha