Learn More
Andwin Scientific Disposable Petri Dishes 100mm x 15mm, Case of 500
SDP
Catalog No. NC0538522
Encompass_Preferred
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a semi-stackable design. These dishes are measured to have a diamter of 100 mm x 15mm
Catalog No. NC0538522 Supplier Andwin Scientific Supplier No. 25384088
The Disposable Petri Dishes are constructed from virgin crystal grade polystyrene that provides optimum flatness and high optical clarity. The mono petri dishes feature a semi-stackable design. These dishes are measured to have a diamter of 100 mm x 15mm