Learn More
Fisherbrand™ Petri Dishes with Clear Lids
Catalog No. 0875714
Encompass_Preferred
Change view
Click to view available options
Closure Type:
Slippable
Stackable
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
08-757-14 -
08-757-13 Slippable
08-757-12 Stackable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 08-757-14 Supplier Fisherbrand™ Supplier No. 42214010000100
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product
Polystyrene, radiation-sterilized dishes are easy to handle with just one hand
Clear tops for distortion-free viewing.
  • Radiation sterilized; sterile if sleeve is unopened
  • Top and bottom edges are squared off (not rounded) to provide a firm, sure grip
  • Bottom is flat so medium spreads evenly over the entire dish bottom
  • Three ventilation ribs allow free air circulation and reduce condensation during incubation
  • Lid skirt is flared just enough so that it can be replaced quickly after streaking
  • Place lid upside down on dish to close off dish from outside environment
Raised-Ridge Petri Dishes
  • Stack high and remain stacked
  • Minimum of slipping, sliding, and resulting scratches
Beveled-Ridge Petri Dishes
  • Ideal for automated filling systems
  • Remove dishes easily from the center of a stack
No-Ridge Petri Dishes
  • Ideal for use in automated systems
Specifications
Disposable Single-use
Material Polystyrene
Sterility Sterile
Packaging Sleeve
Quantity 10/Sleeve
Vent Yes
Height (Metric) 15 mm
Diameter (Metric) 150 mm