Learn More
Sigma Aldrich Fine Chemicals Biosciences Petri dishes polystyrene s
Catalog No. 502901846
Encompass
Note The actual nominal dimensions of these plates possess an inner diameter of 88mm and an outer diameter with lid of 92.4mm.
Catalog No. 50-290-1846 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P5856500EA
Note The actual nominal dimensions of these plates possess an inner diameter of 88mm and an outer diameter with lid of 92.4mm.