Learn More
Sigma Aldrich Fine Chemicals Biosciences Petri dishes polystyrene s
Catalog No. 502900967
Encompass
Designed for easy one hand opening and closing edges of tops and bottoms are squared off to provide a firm sure grip. flare and length of lid skirt allows lid to be removed and replaced easily. Dishes have three venting ribs on the underside of the lid to prevent condensation build up.Standard slippable petri dishClear top and bottom viewing surfacesDisposable (single use) polystyrene petri dish for general microbiological useMolded from medical-grade virgin polystyreneOptically clearRadiation sterilizedContents sterile if sleeve is unopenedVentilation ribs allow for free air circulation and reduce condensation during incubationflared lid skirt and squared corners for easy one-hand operation especially with glovesPlease note Due to these dishes being constructed of polystyrene (not glass) the walls of the dish may be thinner this results in a size difference in the outer diameter of approx 10 mm.
Catalog No. 50-290-0967 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P5606400EA
Designed for easy one hand opening and closing edges of tops and bottoms are squared off to provide a firm sure grip. flare and length of lid skirt allows lid to be removed and replaced easily. Dishes have three venting ribs on the underside of the lid to prevent condensation build up.Standard slippable petri dishClear top and bottom viewing surfacesDisposable (single use) polystyrene petri dish for general microbiological useMolded from medical-grade virgin polystyreneOptically clearRadiation sterilizedContents sterile if sleeve is unopenedVentilation ribs allow for free air circulation and reduce condensation during incubationflared lid skirt and squared corners for easy one-hand operation especially with glovesPlease note Due to these dishes being constructed of polystyrene (not glass) the walls of the dish may be thinner this results in a size difference in the outer diameter of approx 10 mm.