Learn More
Nelson Jameson Company PETRI FILM RAPID YEAST&MLD 50/
Catalog No. NC0974009
Encompass
Petrifilm Rapid Yeast and Mold Count Plates. Yeast and mold results in 48-60 hours. Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes. This plate has been awarded Official Methods of Analysis (OMA) #2014.05 and NF VALIDATION certificate: 3M 01/13-07/14 by AFNOR Certification. Plates use 75% less energy, use 79% less water, produce 75% less greenhouse gases and result in 66% less waste (by weight and volume) compared to competitive agar methods.
Catalog No. NC0974009 Supplier Nelson Jameson Company Supplier No. 4603475 C
Petrifilm Rapid Yeast and Mold Count Plates. Yeast and mold results in 48-60 hours. Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes. This plate has been awarded Official Methods of Analysis (OMA) #2014.05 and NF VALIDATION certificate: 3M 01/13-07/14 by AFNOR Certification. Plates use 75% less energy, use 79% less water, produce 75% less greenhouse gases and result in 66% less waste (by weight and volume) compared to competitive agar methods.