Learn More
Research Products International Corp PetriStickers, 8 Section Pie, 3 Inch Diameter, 36 per Package
SDP
Catalog No. 50998485
Encompass
PetriStickers label petri dishes quickly, neatly and reliably. Convenient stickers are easy to apply to the back of petri dishes. PetriStickers are manufactured with water-resistant ink, substrate and adhesives. Remain firmly attached at temperatures from 4°C to 42°C. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.
Catalog No. 50-998-485 Supplier Research Products International Corp Supplier No. 2470118
PetriStickers label petri dishes quickly, neatly and reliably. Convenient stickers are easy to apply to the back of petri dishes. PetriStickers are manufactured with water-resistant ink, substrate and adhesives. Remain firmly attached at temperatures from 4°C to 42°C. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.