Learn More
Sigma Aldrich Fine Chemicals Biosciences PetriSlides MilliporeR siz
Catalog No. 502901425
Encompass
PetriSlides provide a storage and examination container for 47 mm diameter test filters. Substitute for Petri Dishes. Holds filter securely in place. Transparent cover allows microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.
Catalog No. 50-290-1425 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z355445100EA
PetriSlides provide a storage and examination container for 47 mm diameter test filters. Substitute for Petri Dishes. Holds filter securely in place. Transparent cover allows microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.