Learn More
Research Products International Corp PetriStickers, 32 Square Grid, 3 Inch Diameter, 36 per Package
SDP
Catalog No. 50212508
Encompass
PetriStickers allow for petri dishes to be labeled quickly, neatly and reliably. Made with water-resistant ink, substrate and adhesives, they will remain firmly attached at temperatures from 42° to 4°C. Simply remove the label's protective backing and apply it to the back of a petri dish. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.
Catalog No. 50-212-508 Supplier Research Products International Corp Supplier No. 247010
PetriStickers allow for petri dishes to be labeled quickly, neatly and reliably. Made with water-resistant ink, substrate and adhesives, they will remain firmly attached at temperatures from 42° to 4°C. Simply remove the label's protective backing and apply it to the back of a petri dish. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.