Learn More
Electron Microscopy Sciences Petroff Hausser Counting Chamber 10 µm depth
SDP
Catalog No. 5027811
Encompass_Preferred
New one piece construction ensures durability and accuracy, featuring an improved Neubauer ruling on a single plateau (1/400 sq. mm). Designed for counting bacteria, sperm and blood platelets is offered in a series of cell-depths (10 and 20 microns). Cell-depth is 0.02 mm deep with 1.5 mm thick glass slide allows for the use of dark field microscopy. 2 counting areas
Depth: 10 µm
Catalog No. 50-278-11 Supplier Electron Microscopy Sciences Supplier No. 6351223
New one piece construction ensures durability and accuracy, featuring an improved Neubauer ruling on a single plateau (1/400 sq. mm). Designed for counting bacteria, sperm and blood platelets is offered in a series of cell-depths (10 and 20 microns). Cell-depth is 0.02 mm deep with 1.5 mm thick glass slide allows for the use of dark field microscopy. 2 counting areas
Depth: 10 µm