Learn More
Electron Microscopy Sciences Petrographic Microscope Slide Storage | 100 Slides | Black
SDP
Catalog No. 501931406
Encompass_Preferred
This Petrographic Slide Storage Box is designed for safely storing petrographic slides long term and also for transport. It is made from durable ABS plastic with a hinged lid, secure latch, and foam base.
Catalog No. 50-193-1406 Supplier Electron Microscopy Sciences Supplier No. 7153408
This Petrographic Slide Storage Box is designed for safely storing petrographic slides long term and also for transport. It is made from durable ABS plastic with a hinged lid, secure latch, and foam base.