Learn More
Electron Microscopy Sciences Petrographic Thin Section 100 Slide Storage Box, Blue
SDP
Catalog No. 501931404
Encompass_Preferred
Designed for safely storing petrographic slides long term and also for transport. Made from durable ABS plastic with a hinged lid, secure latch, and foam base.
Color: Blue
Catalog No. 50-193-1404 Supplier Electron Microscopy Sciences Supplier No. 7153406
Designed for safely storing petrographic slides long term and also for transport. Made from durable ABS plastic with a hinged lid, secure latch, and foam base.
Color: Blue