Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Petrol, puriss., 100° to 140°C, Solstice

Catalog No. 60045637 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-637 1 L Glass Bottle
60-045-638 2.5 L Glass Bottle
60-045-640 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-637 Supplier Solstice Supplier No. 245341L
Add to Cart
Add to Cart

Puriss., bp 100-140 °C

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8032-32-4
MDL Number MFCD00081849
Synonym p-tolylmagnesium bromide, magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide

Specifications

Density 0.720 to 0.745g/mL at 20°C
Boiling Point 100°C to 140°C
Flash Point 10°C (50°F)
Packaging Glass Bottle
Quantity 1 L
UN Number UN3295
Grade puriss.
Chemical Name or Material Petrol
Hazard Statement P210-P261-P273-P301 + P310-P331
Precautionary Statement H225-H304-H336-H411
DOTInformation Class 3 - PG 2 - EHS - VP ≤ 110kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR)
EINECSNumber 920-750-0
RTECSNumber OI6180000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.